2H,3H-Naphtho[1,2-b]furan-3-one, tech

Lieferant: Apollo Scientific

OR32552-1G
APOSOR32552-1GEA 1210 EUR
APOSOR32552-1G
2H,3H-Naphtho[1,2-b]furan-3-one, tech
2H,3H-Naphtho[1,2-b]furan-3-one
Formel: C₁₂H₈O₂
Molecular Weight: 184,191 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00975013
CAS-Nummer: 58645-78-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR