2-Chloro-1-methyl-1H-indole-3-carbonitrile, tech

Lieferant: Apollo Scientific

OR32556-1G
APOSOR32556-1GEA 1570 EUR
APOSOR32556-1G
2-Chloro-1-methyl-1H-indole-3-carbonitrile, tech
2-Chloro-1-methyl-1H-indole-3-carbonitrile
Formel: C₁₀H₇ClN₂
Molecular Weight: 190,629 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01815413
CAS-Nummer: 159506-88-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR