1,3-Dimethyl-5-phenoxy-1H-pyrazole-4-carbaldehyde 95%
Lieferant: Apollo Scientific
OR32557-1G
APOSOR32557-1GEA
894
EUR
APOSOR32557-1G
1,3-Dimethyl-5-phenoxy-1H-pyrazole-4-carbaldehyde 95%
1,3-Dimethyl-5-phenoxy-1H-pyrazole-4-carbaldehyde
Formel:
C₁₂H₁₂N₂O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD07698583 CAS-Nummer: 109925-10-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...