Pyrrolidine-1-carboxamidinium acetate

Lieferant: Apollo Scientific
Achtung

OR3256-1G
APOSOR3256-1GEA 0 EUR
APOSOR3256-1G
Pyrrolidine-1-carboxamidinium acetate
Pyrrolidine-1-carboxamidinium acetate
Formel: C₇H₁₅N₃O₂
Molecular Weight: 173.22 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09702434
CAS-Nummer: 91983-81-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR