2-Chloro-4-[(E)-2-(dimethylamino)ethenyl]-6-methylpyridine-3-carbonitrile, tech

Lieferant: Apollo Scientific

OR32560-1G
APOSOR32560-1GEA 1570 EUR
APOSOR32560-1G
2-Chloro-4-[(E)-2-(dimethylamino)ethenyl]-6-methylpyridine-3-carbonitrile, tech
2-Chloro-4-[(E)-2-(dimethylamino)ethenyl]-6-methylpyridine-3-carbonitrile
Formel: C₁₁H₁₂ClN₃
Molecular Weight: 221,686 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01814802
CAS-Nummer: 477888-22-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR