4-[(Pyridin-2-yl)methoxy]benzaldehyde 95%

Lieferant: Apollo Scientific

OR32561-1G OR32561-5G
APOSOR32561-1GEA 0 EUR
APOSOR32561-1G APOSOR32561-5G
4-[(Pyridin-2-yl)methoxy]benzaldehyde 95%
4-[(Pyridin-2-yl)methoxy]benzaldehyde
Formel: C₁₃H₁₁NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00469762
CAS-Nummer: 57748-41-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR