2-[4-(tert-Butoxy)phenyl]acetic acid, tech

Lieferant: Apollo Scientific

OR32562-1G
APOSOR32562-1GEA 538 EUR
APOSOR32562-1G
2-[4-(tert-Butoxy)phenyl]acetic acid, tech
2-[4-(tert-Butoxy)phenyl]acetic acid
Formel: C₁₂H₁₆O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD03411042
CAS-Nummer: 135066-21-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR