Methyl-2-(4-hydroxyphenoxy)acetate 95%
Lieferant: Apollo Scientific
Synonyme:
Methyl 2-(4-hydroxyphenoxy)acetate
OR32563-1G
APOSOR32563-1GEA
700
EUR
APOSOR32563-1G
Methyl-2-(4-hydroxyphenoxy)acetate 95%
Methyl-2-(4-hydroxyphenoxy)acetate
Formel:
C₉H₁₀O₄ Molecular Weight: 182,18 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11046458 CAS-Nummer: 70067-75-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...