(1Z,2E)-3-Phenyl-N-(4H-1,2,4-triazol-4-yl)prop-2-en-1-imine, tech

Lieferant: Apollo Scientific

OR32565-1G
APOSOR32565-1GEA 1570 EUR
APOSOR32565-1G
(1Z,2E)-3-Phenyl-N-(4H-1,2,4-triazol-4-yl)prop-2-en-1-imine, tech
(1Z,2E)-3-Phenyl-N-(4H-1,2,4-triazol-4-yl)prop-2-en-1-imine
Formel: C₁₁H₁₀N₄
Molecular Weight: 198,224 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01326436
CAS-Nummer: 35554-48-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR