4-[(4-Chlorophenyl)methoxy]-1lambda6-thiomorpholine-1,1-dione, tech

Lieferant: Apollo Scientific

OR32567-1G
APOSOR32567-1GEA 1570 EUR
APOSOR32567-1G
4-[(4-Chlorophenyl)methoxy]-1lambda6-thiomorpholine-1,1-dione, tech
4-[(4-Chlorophenyl)methoxy]-1lambda6-thiomorpholine-1,1-dione
Formel: C₁₁H₁₄ClNO₃S
Molecular Weight: 275,752 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02102601
CAS-Nummer: 477889-69-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR