2-(2,5-Dimethyl-1H-pyrrol-1-yl)-5-iodopyridine 95%

Lieferant: Apollo Scientific

OR32568-1G
APOSOR32568-1GEA 671 EUR
APOSOR32568-1G
2-(2,5-Dimethyl-1H-pyrrol-1-yl)-5-iodopyridine 95%
2-(2,5-Dimethyl-1H-pyrrol-1-yl)-5-iodopyridine
Formel: C₁₁H₁₁IN₂
Molecular Weight: 298.123 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02186028
CAS-Nummer: 477889-91-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR