Methyl 2-methyl-5-oxo-1-(2-phenylethyl)-4,5-dihydro-1H-pyrrole-3-carboxylate, tech
Lieferant: Apollo Scientific
OR32570-1G
APOSOR32570-1GEA
1570
EUR
APOSOR32570-1G
Methyl 2-methyl-5-oxo-1-(2-phenylethyl)-4,5-dihydro-1H-pyrrole-3-carboxylate, tech
Methyl 2-methyl-5-oxo-1-(2-phenylethyl)-4,5-dihydro-1H-pyrrole-3-carboxylate
Formel:
C₁₅H₁₇NO₃ Molecular Weight: 259,3 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01893900 CAS-Nummer: 371144-81-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...