1-Cyclopropyl-6-oxo-2-phenylpiperidine-3-carboxylic acid, tech
Lieferant: Apollo Scientific
OR32571-1G
APOSOR32571-1GEA
1570
EUR
APOSOR32571-1G
1-Cyclopropyl-6-oxo-2-phenylpiperidine-3-carboxylic acid, tech
1-Cyclopropyl-6-oxo-2-phenylpiperidine-3-carboxylic acid
Formel:
C₁₅H₁₇NO₃ Molecular Weight: 259,3 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD02571879 CAS-Nummer: 477890-39-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...