6-Hydroxy-2-phenyl-2,3-dihydropyridazin-3-one, tech

Lieferant: Apollo Scientific

OR32572-1G
APOSOR32572-1GEA 1570 EUR
APOSOR32572-1G
6-Hydroxy-2-phenyl-2,3-dihydropyridazin-3-one, tech
6-Hydroxy-2-phenyl-2,3-dihydropyridazin-3-one
Formel: C₁₀H₈N₂O₂
Molecular Weight: 188,183 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00010107
CAS-Nummer: 1698-54-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR