2H,3H,5H,6H-Benzo[h]cinnolin-3-one 95%

Lieferant: Apollo Scientific

OR32574-1G
APOSOR32574-1GEA 874 EUR
APOSOR32574-1G
2H,3H,5H,6H-Benzo[h]cinnolin-3-one 95%
2H,3H,5H,6H-Benzo[h]cinnolin-3-one
Formel: C₁₂H₁₀N₂O
Molecular Weight: 198.221 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01659041
CAS-Nummer: 25823-49-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR