3-(Dimethylamino)-1-(3-methylthiophen-2-yl)but-2-en-1-one, tech

Lieferant: Apollo Scientific

OR32576-1G
APOSOR32576-1GEA 1570 EUR
APOSOR32576-1G
3-(Dimethylamino)-1-(3-methylthiophen-2-yl)but-2-en-1-one, tech
3-(Dimethylamino)-1-(3-methylthiophen-2-yl)but-2-en-1-one
Formel: C₁₁H₁₅NOS
Molecular Weight: 209,308 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03618011
CAS-Nummer: 861209-49-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR