N-(4-Chlorophenyl)-1-oxa-4-azaspiro[4.5]decane-4-carboxamide, tech

Lieferant: Apollo Scientific

OR32577-1G
APOSOR32577-1GEA 1570 EUR
APOSOR32577-1G
N-(4-Chlorophenyl)-1-oxa-4-azaspiro[4.5]decane-4-carboxamide, tech
N-(4-Chlorophenyl)-1-oxa-4-azaspiro[4.5]decane-4-carboxamide
Formel: C₁₅H₁₉ClN₂O₂
Molecular Weight: 294,777 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03618017
CAS-Nummer: 861209-90-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR