2-tert-Butyl-4-(2-hydroxyethyl)-5-methyl-2,3-dihydro-1H-pyrazol-3-one, tech

Lieferant: Apollo Scientific

OR32578-1G
APOSOR32578-1GEA 1570 EUR
APOSOR32578-1G
2-tert-Butyl-4-(2-hydroxyethyl)-5-methyl-2,3-dihydro-1H-pyrazol-3-one, tech
2-tert-Butyl-4-(2-hydroxyethyl)-5-methyl-2,3-dihydro-1H-pyrazol-3-one
Formel: C₁₀H₁₈N₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD03848410
CAS-Nummer: 861210-08-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR