{1-[4-(Aminomethyl)phenyl]piperidin-4-yl}methanol 95%
Lieferant: Apollo Scientific
OR32579-1G
APOSOR32579-1GEA
1330
EUR
APOSOR32579-1G
{1-[4-(Aminomethyl)phenyl]piperidin-4-yl}methanol 95%
{1-[4-(Aminomethyl)phenyl]piperidin-4-yl}methanol
Formel:
C₁₃H₂₀N₂O Molecular Weight: 220,311 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03787065 CAS-Nummer: 861210-09-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...