Methyl 2-(2-cyanophenoxy)acetate, tech
Lieferant: Apollo Scientific
OR32580-1G
APOSOR32580-1GEA
1570
EUR
APOSOR32580-1G
Methyl 2-(2-cyanophenoxy)acetate, tech
Methyl 2-(2-cyanophenoxy)acetate
Formel:
C₁₀H₉NO₃ Molecular Weight: 191,183 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03617964 CAS-Nummer: 34844-79-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...