Methyl 2-{[(1E)-2-cyano-2-(2,4-dichlorophenyl)eth-1-en-1-yl]oxy}acetate, tech

Lieferant: Apollo Scientific

OR32583-1G
APOSOR32583-1GEA 1570 EUR
APOSOR32583-1G
Methyl 2-{[(1E)-2-cyano-2-(2,4-dichlorophenyl)eth-1-en-1-yl]oxy}acetate, tech
Methyl 2-{[(1E)-2-cyano-2-(2,4-dichlorophenyl)eth-1-en-1-yl]oxy}acetate
Formel: C₁₂H₉Cl₂NO₃
Molecular Weight: 286,111 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03787066
CAS-Nummer: 861210-31-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR