(Z)-N-[1-(Pyrazin-2-yl)ethylidene]hydroxylamine 95%

Lieferant: Apollo Scientific

OR32585-1G
APOSOR32585-1GEA 1330 EUR
APOSOR32585-1G
(Z)-N-[1-(Pyrazin-2-yl)ethylidene]hydroxylamine 95%
(Z)-N-[1-(Pyrazin-2-yl)ethylidene]hydroxylamine
Formel: C₆H₇N₃O
Molecular Weight: 137,139 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03618035
CAS-Nummer: 141190-61-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR