(2E)-3-(Dimethylamino)-1-[4-(morpholin-4-yl)phenyl]prop-2-en-1-one, tech

Lieferant: Apollo Scientific

OR32587-1G
APOSOR32587-1GEA 1570 EUR
APOSOR32587-1G
(2E)-3-(Dimethylamino)-1-[4-(morpholin-4-yl)phenyl]prop-2-en-1-one, tech
(2E)-3-(Dimethylamino)-1-[4-(morpholin-4-yl)phenyl]prop-2-en-1-one
Formel: C₁₅H₂₀N₂O₂
Molecular Weight: 260.332 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03617857
CAS-Nummer: 1345865-34-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR