1-Methyl-1H-pyrazolo[3,4-b]pyridin-3-ylamine 97%

Lieferant: Apollo Scientific

OR32589-5G
APOSOR32589-5GEA 142 EUR
APOSOR32589-5G
1-Methyl-1H-pyrazolo[3,4-b]pyridin-3-ylamine 97%
1-Methyl-1H-pyrazolo[3,4-b]pyridin-3-ylamine
Formel: C₇H₈N₄
Lagertemperatur: Raumtemperatur
MDL: MFCD04117796
CAS-Nummer: 72583-83-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR