4-Ureidobenzeneboronic acid pinacol ester 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-(Carbamoylamino)benzeneboronic acid, pinacol ester, 1-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)urea

OR3259-1G
APOSOR3259-1GEA 133 EUR
APOSOR3259-1G
4-Ureidobenzeneboronic acid pinacol ester 98%
4-Ureidobenzeneboronic acid pinacol ester
Formel: C₁₃H₁₉BN₂O₃
Molecular Weight: 262.12 g/mol
Schmelzpunkt: 124…128 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06795683
CAS-Nummer: 877134-77-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR