2-Chloro-5-[(2-methylpiperidin-1-yl)methyl]pyridine 95%

Lieferant: Apollo Scientific

OR32592-1G
APOSOR32592-1GEA 671 EUR
APOSOR32592-1G
2-Chloro-5-[(2-methylpiperidin-1-yl)methyl]pyridine 95%
2-Chloro-5-[(2-methylpiperidin-1-yl)methyl]pyridine
Formel: C₁₂H₁₇ClN₂
Molecular Weight: 224,73 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD05256124
CAS-Nummer: 861211-51-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR