2-Chloro-5-[(1H-1,2,4-triazol-1-yl)methyl]pyridine 95%

Lieferant: Apollo Scientific

OR32593-1G
APOSOR32593-1GEA 671 EUR
APOSOR32593-1G
2-Chloro-5-[(1H-1,2,4-triazol-1-yl)methyl]pyridine 95%
2-Chloro-5-[(1H-1,2,4-triazol-1-yl)methyl]pyridine
Formel: C₈H₇ClN₄
Molecular Weight: 194,621 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04125022
CAS-Nummer: 861211-53-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR