3-{[(6-Chloropyridin-3-yl)methyl](methyl)amino}propanenitrile, tech

Lieferant: Apollo Scientific

OR32594-1G
APOSOR32594-1GEA 1570 EUR
APOSOR32594-1G
3-{[(6-Chloropyridin-3-yl)methyl](methyl)amino}propanenitrile, tech
3-{[(6-Chloropyridin-3-yl)methyl](methyl)amino}propanenitrile
Formel: C₁₀H₁₂ClN₃
Molecular Weight: 209,675 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04125025
CAS-Nummer: 861211-56-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR