1-tert-Butyl-6-(4-chlorophenyl)piperidine-2,4-dione, tech
Lieferant: Apollo Scientific
OR32601-1G
APOSOR32601-1GEA
1570
EUR
APOSOR32601-1G
1-tert-Butyl-6-(4-chlorophenyl)piperidine-2,4-dione, tech
1-tert-Butyl-6-(4-chlorophenyl)piperidine-2,4-dione
Formel:
C₁₅H₁₈ClNO₂ Molecular Weight: 279,762 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01936197 CAS-Nummer: 400079-95-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...