7-Chloro-2H-[1,4]benzothiazin-3(4H)-one 95%

Lieferant: Apollo Scientific

OR32602-5G
APOSOR32602-5GEA 270 EUR
APOSOR32602-5G
7-Chloro-2H-[1,4]benzothiazin-3(4H)-one 95%
7-Chloro-2H-[1,4]benzothiazin-3(4H)-one
Formel: C₈H₆ClNOS
Molecular Weight: 199.67 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00127344
CAS-Nummer: 5333-05-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR