3-Methyl-1-(1,4,5,6-tetrahydropyrimidin-2-yl)-4,5-dihydro-1H-pyrazol-5-one, tech

Lieferant: Apollo Scientific

OR32603-1G
APOSOR32603-1GEA 1570 EUR
APOSOR32603-1G
3-Methyl-1-(1,4,5,6-tetrahydropyrimidin-2-yl)-4,5-dihydro-1H-pyrazol-5-one, tech
3-Methyl-1-(1,4,5,6-tetrahydropyrimidin-2-yl)-4,5-dihydro-1H-pyrazol-5-one
Formel: C₈H₁₂N₄O
Molecular Weight: 180,207 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00140900
CAS-Nummer: 338402-58-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR