3-[(4-Hydroxyphenyl)methyl]pentane-2,4-dione, tech

Lieferant: Apollo Scientific

OR32604-1G
APOSOR32604-1GEA 1570 EUR
APOSOR32604-1G
3-[(4-Hydroxyphenyl)methyl]pentane-2,4-dione, tech
3-[(4-Hydroxyphenyl)methyl]pentane-2,4-dione
Formel: C₁₂H₁₄O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD00138768
CAS-Nummer: 15451-07-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR