2-(4-Chlorophenyl)-3-(dimethylamino)acrylonitrile 95%

Lieferant: Apollo Scientific

Synonyme: (2Z)-2-(4-Chlorophenyl)-3-(dimethylamino)prop-2-enenitrile

OR32606-1G
APOSOR32606-1GEA 219 EUR
APOSOR32606-1G
2-(4-Chlorophenyl)-3-(dimethylamino)acrylonitrile 95%
2-(4-Chlorophenyl)-3-(dimethylamino)acrylonitrile
Formel: C₁₁H₁₁ClN₂
Molecular Weight: 206.68 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00138415
CAS-Nummer: 40373-88-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR