N,N-Dimethyl-4-(2-phenylacetyl)-1H-pyrrole-2-carboxamide, tech
Lieferant: Apollo Scientific
OR32609-1G
APOSOR32609-1GEA
1570
EUR
APOSOR32609-1G
N,N-Dimethyl-4-(2-phenylacetyl)-1H-pyrrole-2-carboxamide, tech
N,N-Dimethyl-4-(2-phenylacetyl)-1H-pyrrole-2-carboxamide
Formel:
C₁₅H₁₆N₂O₂ Molecular Weight: 256,3 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00141860 CAS-Nummer: 338403-34-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...