1-[(3,4-Dichlorophenyl)methoxy]-3-[(1E)-(dimethylamino)methylidene]urea, tech
Lieferant: Apollo Scientific
OR32610-1G
APOSOR32610-1GEA
1570
EUR
APOSOR32610-1G
1-[(3,4-Dichlorophenyl)methoxy]-3-[(1E)-(dimethylamino)methylidene]urea, tech
1-[(3,4-Dichlorophenyl)methoxy]-3-[(1E)-(dimethylamino)methylidene]urea
Formel:
C₁₁H₁₃Cl₂N₃O₂ Molecular Weight: 290,146 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00139685 CAS-Nummer: 338404-01-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...