4-(2-Chlorobenzoyl)-N,N-dimethyl-1H-pyrrole-2-carboxamide, tech
Lieferant: Apollo Scientific
OR32611-1G
APOSOR32611-1GEA
1570
EUR
APOSOR32611-1G
4-(2-Chlorobenzoyl)-N,N-dimethyl-1H-pyrrole-2-carboxamide, tech
4-(2-Chlorobenzoyl)-N,N-dimethyl-1H-pyrrole-2-carboxamide
Formel:
C₁₄H₁₃ClN₂O₂ Molecular Weight: 276,718 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00141879 CAS-Nummer: 338404-09-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...