2,2-Bis(hydroxymethyl)-1,2,3,4-tetrahydronaphthalen-1-one, tech

Lieferant: Apollo Scientific

OR32613-1G
APOSOR32613-1GEA 1570 EUR
APOSOR32613-1G
2,2-Bis(hydroxymethyl)-1,2,3,4-tetrahydronaphthalen-1-one, tech
2,2-Bis(hydroxymethyl)-1,2,3,4-tetrahydronaphthalen-1-one
Formel: C₁₂H₁₄O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD00139856
CAS-Nummer: 15121-83-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR