4-Chloro-1-(4-chlorophenyl)-6-oxo-1,6-dihydropyridazine-3-carbonitrile, tech
Lieferant: Apollo Scientific
OR32614-1G
APOSOR32614-1GEA
1570
EUR
APOSOR32614-1G
4-Chloro-1-(4-chlorophenyl)-6-oxo-1,6-dihydropyridazine-3-carbonitrile, tech
4-Chloro-1-(4-chlorophenyl)-6-oxo-1,6-dihydropyridazine-3-carbonitrile
Formel:
C₁₁H₅Cl₂N₃O Molecular Weight: 266,083 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00141174 CAS-Nummer: 338405-10-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...