Methyl 4-chloro-1-(4-chlorophenyl)-6-oxo-1,6-dihydropyridazine-3-carboxylate, tech
Lieferant: Apollo Scientific
OR32615-1G
APOSOR32615-1GEA
1570
EUR
APOSOR32615-1G
Methyl 4-chloro-1-(4-chlorophenyl)-6-oxo-1,6-dihydropyridazine-3-carboxylate, tech
Methyl 4-chloro-1-(4-chlorophenyl)-6-oxo-1,6-dihydropyridazine-3-carboxylate
Formel:
C₁₂H₈Cl₂N₂O₃ Molecular Weight: 299,11 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00141184 CAS-Nummer: 129109-19-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...