2-[(Quinoxalin-2-yl)amino]ethan-1-ol, tech
Lieferant: Apollo Scientific
OR32616-1G
APOSOR32616-1GEA
1570
EUR
APOSOR32616-1G
2-[(Quinoxalin-2-yl)amino]ethan-1-ol, tech
2-[(Quinoxalin-2-yl)amino]ethan-1-ol
Formel:
C₁₀H₁₁N₃O Molecular Weight: 189,214 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00243663 CAS-Nummer: 30466-69-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...