[1,2,4]Triazolo[1,5-a]pyridin-8-amine 95%

Lieferant: Apollo Scientific

OR32617-1G
APOSOR32617-1GEA 671 EUR
APOSOR32617-1G
[1,2,4]Triazolo[1,5-a]pyridin-8-amine 95%
[1,2,4]Triazolo[1,5-a]pyridin-8-amine
Formel: C₆H₆N₄
Lagertemperatur: Raumtemperatur
MDL: MFCD01316370
CAS-Nummer: 31052-95-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR