(2-Methyl-3-phenyltetrahydro-5-isoxazolyl)methanol 95%

Lieferant: Apollo Scientific

OR32618-1G
APOSOR32618-1GEA 671 EUR
APOSOR32618-1G
(2-Methyl-3-phenyltetrahydro-5-isoxazolyl)methanol 95%
(2-Methyl-3-phenyltetrahydro-5-isoxazolyl)methanol
Formel: C₁₁H₁₅NO₂
Molecular Weight: 193,25 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00171848
CAS-Nummer: 19345-02-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR