Methyl (2E)-3-amino-4-(4-chlorophenyl)but-2-enoate, tech
Lieferant: Apollo Scientific
OR32619-1G
APOSOR32619-1GEA
1210
EUR
APOSOR32619-1G
Methyl (2E)-3-amino-4-(4-chlorophenyl)but-2-enoate, tech
Methyl (2E)-3-amino-4-(4-chlorophenyl)but-2-enoate
Formel:
C₁₁H₁₂ClNO₂ Molecular Weight: 225,671 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00215505 CAS-Nummer: 338406-63-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...