2-[(4-Chlorobenzyl)oxy]-1-ethanol 97%

Lieferant: Apollo Scientific

OR32621-1G
APOSOR32621-1GEA 154 EUR
APOSOR32621-1G
2-[(4-Chlorobenzyl)oxy]-1-ethanol 97%
2-[(4-Chlorobenzyl)oxy]-1-ethanol
Formel: C₉H₁₁ClO₂
Molecular Weight: 186.64 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00172032
CAS-Nummer: 1200-15-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR