1-Chloro-4-[(2-chloroethoxy)methyl]benzene 95%

Lieferant: Apollo Scientific

OR32622-1G
APOSOR32622-1GEA 1330 EUR
APOSOR32622-1G
1-Chloro-4-[(2-chloroethoxy)methyl]benzene 95%
1-Chloro-4-[(2-chloroethoxy)methyl]benzene
Formel: C₉H₁₀Cl₂O
Molecular Weight: 205,081 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00172034
CAS-Nummer: 29904-52-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR