1,5,6-Trimethyl-1H-1,3-benzodiazol-4-amine, tech

Lieferant: Apollo Scientific

OR32625-1G
APOSOR32625-1GEA 1570 EUR
APOSOR32625-1G
1,5,6-Trimethyl-1H-1,3-benzodiazol-4-amine, tech
1,5,6-Trimethyl-1H-1,3-benzodiazol-4-amine
Formel: C₁₀H₁₃N₃
Molecular Weight: 175.23 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00231368
CAS-Nummer: 338410-75-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR