1-Phenyl-1H-imidazole-2-thiol 95%
Lieferant: Apollo Scientific
OR32630-5G
APOSOR32630-5GEA
0
EUR
APOSOR32630-5G
1-Phenyl-1H-imidazole-2-thiol 95%
1-Phenyl-1H-imidazole-2-thiol
Formel:
C₉H₈N₂S Molecular Weight: 176.24 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00127929 CAS-Nummer: 17452-09-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...