4-Methoxy-2-(1H-pyrrol-1-yl)pyridine-3-carbonitrile, tech
Lieferant: Apollo Scientific
OR32631-1G
APOSOR32631-1GEA
1570
EUR
APOSOR32631-1G
4-Methoxy-2-(1H-pyrrol-1-yl)pyridine-3-carbonitrile, tech
4-Methoxy-2-(1H-pyrrol-1-yl)pyridine-3-carbonitrile
Formel:
C₁₁H₉N₃O Molecular Weight: 199,209 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00975449 CAS-Nummer: 338412-19-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...