2-Chloro-4,6-dimethyl-3-(methylsulphonyl)pyridine 95%

Lieferant: Apollo Scientific

Synonyme: 2-Chloro-3-methanesulphonyl-4,6-dimethylpyridine

OR32632-1G
APOSOR32632-1GEA 538 EUR
APOSOR32632-1G
2-Chloro-4,6-dimethyl-3-(methylsulphonyl)pyridine 95%
2-Chloro-4,6-dimethyl-3-(methylsulphonyl)pyridine
Formel: C₈H₁₀ClNO₂S
Molecular Weight: 219,69 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08056620
CAS-Nummer: 886361-59-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR