5-Chloro-1-methyl-3-phenyl-1H-pyrazole-4-carboxylic acid 95%

Lieferant: Apollo Scientific

OR32633-1G
APOSOR32633-1GEA 458 EUR
APOSOR32633-1G
5-Chloro-1-methyl-3-phenyl-1H-pyrazole-4-carboxylic acid 95%
5-Chloro-1-methyl-3-phenyl-1H-pyrazole-4-carboxylic acid
Formel: C₁₁H₉ClN₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD01314939
CAS-Nummer: 191419-14-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR